missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD155/PVR Rabbit anti-Mouse, Clone: 305, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90501-100UL
This item is not returnable.
View return policy
Description
CD155/PVR Monoclonal specifically detects CD155/PVR in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| CD155/PVR | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunofluorescence | |
| 305 | |
| Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:1000 | |
| CD155, CD155 antigen, HVED, NECL5, NECL-5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVSFLJ25946, Tage4 | |
| Recombinant Mouse CD155/PVR protein | |
| 100 μg | |
| Virology Bacteria and Parasites | |
| 5817 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction