missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD155/PVR Antibody (D171), Novus Biologicals™
Mouse Monoclonal Antibody
£149.00
Specifications
| Antigen | CD155/PVR |
|---|---|
| Clone | D171 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1:10 - 1:100, Flow Cytometry 2 μg, Immunocytochemistry/Immunofluorescence 1:10 - 1:100 |
| Applications | Western Blot |
Description
CD155/PVR Monoclonal specifically detects CD155/PVR in Human, Mouse, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
| CD155/PVR | |
| 0.2 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Virology Bacteria and Parasites | |
| CD155, CD155 antigen, HVED, Necl-5, NECL5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVSFLJ25946, Tage4 | |
| PVR | |
| IgG1 | |
| Protein G purified | |
| Polio Virus Receptor (D171) |
| D171 | |
| Western Blot 1:10 - 1:100, Flow Cytometry 2 μg, Immunocytochemistry/Immunofluorescence 1:10 - 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 5817 | |
| HeLa cells | |
| Primary | |
| Store at 4°C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title