missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD155/PVR Antibody (D171), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-1241-100ul
This item is not returnable.
View return policy
Description
CD155/PVR Monoclonal specifically detects CD155/PVR in Human, Mouse, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| CD155/PVR | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1:10 - 1:100, Flow Cytometry 2 μg, Immunocytochemistry/Immunofluorescence 1:10 - 1:100 | |
| PVR | |
| HeLa cells | |
| 100 μL | |
| Virology Bacteria and Parasites | |
| 5817 | |
| Human | |
| Purified |
| Western Blot | |
| D171 | |
| Unconjugated | |
| CD155, CD155 antigen, HVED, Necl-5, NECL5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVSFLJ25946, Tage4 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Polio Virus Receptor (D171) | |
| Store at 4°C. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction