Learn More
CD154 Armenian Hamster, FITC, Clone: MR1, enQuireBio™
Armenian Hamster Monoclonal Antibody
Brand:  EnquireBio QAB88-F-25ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15962598
  • £62.45 / 25µg
Estimated Shipment: 19-06-2024
to see stock.
Specifications
CD154
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Cd40lg
Protein A/G Purified
RUO
21947
IgG
Flow Cytometry
MR1
FITC
NP_035746
Armenian Hamster
25 μg
Primary
Store at 2-4°C.
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
Certificats