missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD154 Armenian Hamster, FITC, Clone: MR1, enQuireBio™
Armenian Hamster Monoclonal Antibody
£62.45 - £155.00
Specifications
Antigen | CD154 |
---|---|
Clone | MR1 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15962598
|
EnquireBio
QAB88-F-25UG |
25 μg |
£62.45
25µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15952598
|
EnquireBio
QAB88-F-100UG |
100 μg |
£155.00
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD154 | |
0.5mg/mL | |
Monoclonal | |
Armenian Hamster | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
21947 | |
IgG | |
Protein A/G Purified |
MR1 | |
Flow Cytometry | |
FITC | |
RUO | |
NP_035746 | |
Cd40lg | |
Primary | |
Store at 2-4°C. |