missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD151 Mouse anti-Human, Clone: B-E31, Novus Biologicals™
Mouse Monoclonal Antibody
£305.00 - £572.00
Specifications
| Antigen | CD151 |
|---|---|
| Clone | B-E31 |
| Concentration | 1 mg/ml |
| Dilution | Flow Cytometry |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18390224
|
Novus Biologicals
NBP3-18100-200UL |
200 μg |
£305.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18304687
|
Novus Biologicals
NBP3-18100-500UL |
500 μg |
£572.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD151 Monoclonal antibody specifically detects CD151 in Human samples. It is validated for Flow CytometrySpecifications
| CD151 | |
| 1 mg/ml | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD151 antigen (Raph blood group), CD151 antigenplatelet surface glycoprotein gp27, CD151 molecule (Raph blood group), GP27, Membrane glycoprotein SFA-1, MER2, PETA3, PETA-3SFA-1, platelet-endothelial cell tetraspan antigen 3, Platelet-endothelial tetraspan antigen 3, RAPH, SFA1, tetraspanin-24, tspan-24, TSPAN24hemidesmosomal tetraspanin CD151 | |
| Eahy 926 cell line. Hybridoma: Myeloma X63/AG.8653 x Balb/c spleen cell | |
| Primary | |
| Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles. |
| B-E31 | |
| Flow Cytometry | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS. Sterile-filtered through 0.22 um. | |
| 977 | |
| IgG1 κ | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title