missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD151 Mouse anti-Human, Clone: B-E31, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18100-500UL
This item is not returnable.
View return policy
Description
CD151 Monoclonal antibody specifically detects CD151 in Human samples. It is validated for Flow Cytometry
Specifications
| CD151 | |
| Monoclonal | |
| 1 mg/ml | |
| Flow Cytometry | |
| CD151 antigen (Raph blood group), CD151 antigenplatelet surface glycoprotein gp27, CD151 molecule (Raph blood group), GP27, Membrane glycoprotein SFA-1, MER2, PETA3, PETA-3SFA-1, platelet-endothelial cell tetraspan antigen 3, Platelet-endothelial tetraspan antigen 3, RAPH, SFA1, tetraspanin-24, tspan-24, TSPAN24hemidesmosomal tetraspanin CD151 | |
| Eahy 926 cell line. Hybridoma: Myeloma X63/AG.8653 x Balb/c spleen cell | |
| 500 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| B-E31 | |
| Unconjugated | |
| PBS. Sterile-filtered through 0.22 um. | |
| Mouse | |
| Affinity purified | |
| RUO | |
| 977 | |
| Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction