Learn More
CD150 Monoclonal Antibody (9D1), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£7770.00 - £12950.00
Specifications
| Antigen | CD150 |
|---|---|
| Clone | 9D1 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, Functional Assay |
| Classification | Monoclonal |
Description
The 9D1 MAb reacts with mouse CD150, an ∽70 kDa transmembrane glycoprotein also known as Signaling Lymphocyte Activation Molecule (SLAM). CD150 is expressed by T (especially TH1) and B cells and its expression is rapidly upregulated on these cells upon activation. Immature thymocytes and dendritic cells also express this antigen. Signaling through SLAM in T cells induces proliferation and augmentation of the interferon-gamma response. Furthermore, SLAM is thought to play a role in adhesion between the T cell and antigen-presenting cell. 9D1 is reported to be an activating antibody. Mouse hematopoietic stem cells (HSC) can be identified using SLAM family markers, such as CD150+CD244-CD48-. For this application we recommend the use of antibody clone mShad150 (cat. 12-1502).
CD150 Monoclonal antibody specifically detects CD150 in Mouse samples. It is validated for Flow Cytometry, Functional AssaySpecifications
| CD150 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| 4933415F16; AA177906; CD150; CD150 antigen; CDw150; ESTM51; IPO-3; RGD1560634; signaling lymphocyte activation molecule; signaling lymphocytic activation molecule; signaling lymphocytic activation molecule family member 1; Slam; SLAM family member 1; SLAMF1 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| 9D1 | |
| Flow Cytometry, Functional Assay | |
| Functional Grade | |
| Rat | |
| Mouse | |
| Q9QUM4 | |
| 27218 | |
| Primary | |
| 4° C | |
| Slamf1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.