Learn More
CD150 Monoclonal Antibody (9D1), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand:  Affymetrix eBioscience 16-1501-025
 View more versions of this product
Product Code. 17191152
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
The 9D1 MAb reacts with mouse CD150, an ∽70 kDa transmembrane glycoprotein also known as Signaling Lymphocyte Activation Molecule (SLAM). CD150 is expressed by T (especially TH1) and B cells and its expression is rapidly upregulated on these cells upon activation. Immature thymocytes and dendritic cells also express this antigen. Signaling through SLAM in T cells induces proliferation and augmentation of the interferon-gamma response. Furthermore, SLAM is thought to play a role in adhesion between the T cell and antigen-presenting cell. 9D1 is reported to be an activating antibody. Mouse hematopoietic stem cells (HSC) can be identified using SLAM family markers, such as CD150+CD244-CD48-. For this application we recommend the use of antibody clone mShad150 (cat. 12-1502).
CD150 Monoclonal antibody specifically detects CD150 in Mouse samples. It is validated for Flow Cytometry, Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD150
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.2
Q9QUM4
Rat
25 mg
Primary
Mouse
Antibody
IgG1 κ
Flow Cytometry, Functional Assay
9D1
Functional Grade
Slamf1
4933415F16; AA177906; CD150; CD150 antigen; CDw150; ESTM51; IPO-3; RGD1560634; signaling lymphocyte activation molecule; signaling lymphocytic activation molecule; signaling lymphocytic activation molecule family member 1; Slam; SLAM family member 1; SLAMF1
Affinity chromatography
RUO
27218
4° C