missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD138/Syndecan-1 Mouse anti-Human, Unconjugated, Clone: 4F3A8, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CD138/Syndecan-1 |
|---|---|
| Clone | 4F3A8 |
| Concentration | 1000 μg/mL |
| Applications | Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16869625
|
Proteintech
67155-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16859625
|
Proteintech
67155-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD138/Syndecan-1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD138/Syndecan-1. This antibody recognizes human antigen. The CD138/Syndecan-1 antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
| CD138/Syndecan-1 | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 6382 | |
| Primary |
| 4F3A8 | |
| Immunohistochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Human | |
| IgG2a |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title