Learn More
CD138/Syndecan-1 Mouse anti-Human, Unconjugated, Clone: 4F3A8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67155-1-IG-20UL
Description
The CD138/Syndecan-1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD138/Syndecan-1. This antibody recognizes human antigen. The CD138/Syndecan-1 antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
| CD138/Syndecan-1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Immunohistochemistry, Immunofluorescence, ELISA | |
| 4F3A8 | |
| Unconjugated | |
| 20 μL | |
| 6382 | |
| IgG2a |