missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD133 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£159.00 - £358.00
Specifications
| Antigen | CD133 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 2 ug/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18199558
|
Novus Biologicals
NBP2-44250SS |
0.025 mg |
£159.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18153399
|
Novus Biologicals
NBP2-44250 |
0.1 mg |
£358.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifically detects CD133 in Mouse samples, and it is validated for Immunocytochemistry, Immunofluorescence, Western Blotting
CD133 Polyclonal specifically detects CD133 in Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| CD133 | |
| Western Blot 2 ug/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Cellular Markers, Core ESC Like Genes, Hematopoietic Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers | |
| PBS, 30% Glycerol with 0.02% Sodium Azide | |
| 8842 | |
| A synthetic peptide made to an internal portion of the mouse CD133 protein (between residues 650-750) [UniProt O54990] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Mouse | |
| AC133 Stargardt disease 4 (autosomal dominant), CD133, CD133 retinal 2, prominin (mouse)-like 1, prominin 1 | |
| PROM1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title