missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD133 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44250
This item is not returnable.
View return policy
Description
Specifically detects CD133 in Mouse samples, and it is validated for Immunocytochemistry, Immunofluorescence, Western Blotting
CD133 Polyclonal specifically detects CD133 in Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| CD133 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 30% Glycerol with 0.02% Sodium Azide | |
| PROM1 | |
| A synthetic peptide made to an internal portion of the mouse CD133 protein (between residues 650-750) [UniProt O54990] | |
| 0.1 mg | |
| Cellular Markers, Core ESC Like Genes, Hematopoietic Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers | |
| 8842 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1.0 mg/mL | |
| Western Blot 2 ug/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| AC133 Stargardt disease 4 (autosomal dominant), CD133, CD133 retinal 2, prominin (mouse)-like 1, prominin 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction