Learn More
CD132 Mouse anti-Human, PE, Clone: AG184, BD
Mouse Monoclonal Antibody
£127.68 - £340.86
Specifications
| Antigen | CD132 |
|---|---|
| Clone | AG184 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15841249
|
BD Biosciences
561699 |
25 μg |
£134.00 £127.68 / Each Save £6.32 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15848518
|
BD Biosciences
555900 |
0.2 mg |
£356.00 £340.86 / Each Save £15.14 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TheAG184 monoclonal antibody specifically binds to the 65-70 kDa common γ subunit (γc) that is shared by the IL-2, IL-4, IL-7, IL-9, IL-15 and IL-21 receptor complexes. The γc receptor is a type 1 transmembrane glycoprotein that is constitutively expressed by most peripheral T and B lymphocytes, NK cells, monocytes and granulocytes. The cytoplasmic domain of the γc chain plays an important role in cytokine-mediated signal transduction. By immunofluorescent staining and flow cytometric analyses, the AG184 antibody has been shown to specifically recognize human γc expressed by cell lines including human γc gene-transfected cell lines which are known to express the human γc chain. The AG184 antibody can bind the γc chain in the receptors complexed with IL-2, IL-4, or IL-7, indicating that the antibody recognizes an epitope which is distinct from the cytokine binding site of the γc chain.
Specifications
| CD132 | |
| 0.2mg/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG1 κ | |
| Affinity Purified | |
| IL2RG; IL-2RG; IL-2Rγ; Common gamma chain; γc; CIDX; SCIDX; SCIDX1; IMD4 |
| AG184 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.