missing translation for 'onlineSavingsMsg'
Learn More

CD132 Mouse anti-Human, PE, Clone: AG184, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 555900

 View more versions of this product

Product Code. 15848518

  • ONLINE EXCLUSIVE PRICE
    £356.00
    £340.86 / Each
    Save £15.14
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TheAG184 monoclonal antibody specifically binds to the 65-70 kDa common γ subunit (γc) that is shared by the IL-2, IL-4, IL-7, IL-9, IL-15 and IL-21 receptor complexes. The γc receptor is a type 1 transmembrane glycoprotein that is constitutively expressed by most peripheral T and B lymphocytes, NK cells, monocytes and granulocytes. The cytoplasmic domain of the γc chain plays an important role in cytokine-mediated signal transduction. By immunofluorescent staining and flow cytometric analyses, the AG184 antibody has been shown to specifically recognize human γc expressed by cell lines including human γc gene-transfected cell lines which are known to express the human γc chain. The AG184 antibody can bind the γc chain in the receptors complexed with IL-2, IL-4, or IL-7, indicating that the antibody recognizes an epitope which is distinct from the cytokine binding site of the γc chain.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD132
Monoclonal
0.2mg/mL
IL2RG; IL-2RG; IL-2Rγ; Common gamma chain; γc; CIDX; SCIDX; SCIDX1; IMD4
Mouse
0.2 mg
Immunology
Human
IgG1 κ
Flow Cytometry
AG184
PE
Aqueous buffered solution containing ≤0.09% sodium azide.
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.