missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Mouse, Clone: ITGAX/1243, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £453.00
Specifications
Antigen | CD11c |
---|---|
Clone | ITGAX/1243 |
Applications | Immunohistochemistry |
Conjugate | Unconjugated |
Host Species | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15960288
|
EnquireBio
3687-MSM3-P1 |
100 μg |
£453.00
100µg |
Estimated Shipment: 21-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15950288
|
EnquireBio
3687-MSM3-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
Specifications
CD11c | |
Immunohistochemistry | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
3687 | |
IgG2b κ | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
145kDa |
ITGAX/1243 | |
Unconjugated | |
RUO | |
XP_011544154 | |
Recombinant human ITGAX protein fragment (aa 637-827) | |
Protein A/G Purified | |
Cell surface |