missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Mouse, Clone: ITGAX/1243, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3687-MSM3-P0
Additional Details : Weight : 0.01000kg
Specifications
CD11c | |
Cell surface | |
ITGAX/1243 | |
Unconjugated | |
XP_011544154 | |
Recombinant human ITGAX protein fragment (aa 637-827) | |
Human dendritic cells. Human lymph nodes and tonsils. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
Mouse | |
145kDa | |
Protein A/G Purified | |
RUO | |
3687 | |
IgG2b κ |