Learn More
CD11c Armenian Hamster, FITC, Clone: N418, enQuireBio™
Armenian Hamster Monoclonal Antibody
Brand:  EnquireBio QAB23-F-500ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15951698
  • £191.00 / 500µg
Estimated Shipment: 20-06-2024
to see stock.
Specifications
CD11c
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Itgax
Protein A/G Purified
RUO
16411
IgG
Flow Cytometry
N418
FITC
NP_067309
Armenian Hamster
500 μg
Primary
Store at 2-4°C.