missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Armenian Hamster, FITC, Clone: N418, enQuireBio™
Armenian Hamster Monoclonal Antibody
£62.45 - £191.00
Specifications
Antigen | CD11c |
---|---|
Clone | N418 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15941698
|
EnquireBio
QAB23-F-25UG |
25 μg |
£62.45
25µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15931698
|
EnquireBio
QAB23-F-100UG |
100 μg |
£109.00
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15951698
|
EnquireBio
QAB23-F-500UG |
500 μg |
£191.00
500µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD11c | |
0.5mg/mL | |
Monoclonal | |
Armenian Hamster | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
16411 | |
IgG | |
Protein A/G Purified |
N418 | |
Flow Cytometry | |
FITC | |
RUO | |
NP_067309 | |
Itgax | |
Primary | |
Store at 2-4°C. |