missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Antibody (AP-MAB0814) - Azide and BSA Free, Novus Biologicals™
Armenian Hamster Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-06651-0.1mg
This item is not returnable.
View return policy
Description
CD11c Monoclonal specifically detects CD11c in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, CyTOF-ready.
Specifications
| CD11c | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen, CyTOF-ready | |
| 95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, SLEB6 | |
| Armenian Hamster | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| The was selected for its ability to detect mouse dendritic cells, NK cells, intestinal intraepithelial lymphocytes (IEL), and some activated T cells. | |
| Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles. | |
| IgG |
| Flow Cytometry | |
| AP-MAB0814 | |
| Unconjugated | |
| PBS with No Preservative | |
| ITGAX | |
| Mouse CD11c+ T Cells | |
| 0.1 mg | |
| Dendritic Cell Markers, Signal Transduction | |
| 3687 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction