missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Antibody (AP-MAB0814) - Azide and BSA Free, Novus Biologicals™
Armenian Hamster Monoclonal Antibody has been used in 3 publications
£150.00
Specifications
| Antigen | CD11c |
|---|---|
| Clone | AP-MAB0814 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen, CyTOF-ready |
| Applications | Flow Cytometry |
Description
CD11c Monoclonal specifically detects CD11c in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, CyTOF-ready.Specifications
| CD11c | |
| 1 mg/mL | |
| Flow Cytometry | |
| Unconjugated | |
| Armenian Hamster | |
| Dendritic Cell Markers, Signal Transduction | |
| PBS with No Preservative | |
| 3687 | |
| Mouse CD11c+ T Cells | |
| Primary | |
| Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles. |
| AP-MAB0814 | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| 95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, SLEB6 | |
| ITGAX | |
| IgG | |
| Protein A or G purified | |
| The was selected for its ability to detect mouse dendritic cells, NK cells, intestinal intraepithelial lymphocytes (IEL), and some activated T cells. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title