Learn More
Invitrogen™ CD105 (Endoglin) Monoclonal Antibody (SN6), Functional Grade, eBioscience™

Mouse Monoclonal Antibody
£204.00
Specifications
| Antigen | CD105 (Endoglin) |
|---|---|
| Clone | SN6 |
| Concentration | 1 mg/mL |
| Content And Storage | 4°C |
| Applications | Functional Assay |
Description
Description: The SN6 monoclonal antibody reacts with human CD105, also known as Endoglin. CD105, an approximately 90 kDa disulfide-linked homodimer is expressed by vascular endothelial cells and some bone marrow cells and activated macrophages. It is suggested that CD105 functions in adhesion and embryonic angiogenesis. Applications Reported: This SN6 antibody has been reported to have functional activity. Applications Tested: This SN6 antibody has been tested by flow cytometric analysis of U937 cell line. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
CD105 or Endoglin is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists on an O- and N-glycosylated homodimer. Up regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 binds to TGF beta 1 and 3 with high affinity but not to TGF beta 2.Specifications
| CD105 (Endoglin) | |
| 1 mg/mL | |
| Functional Assay | |
| Functional Grade | |
| Mouse | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| P17813 | |
| 2022 | |
| Primary | |
| Affinity chromatography |
| SN6 | |
| 4°C | |
| Monoclonal | |
| Liquid | |
| IgG1 κ | |
| Human | |
| ENG | |
| AI528660; AI662476; CD105; CD105 antigen; cell surface MJ7/18 antigen; Edg; END; Endo; Endoglin; endoglin (Osler-Rendu-Weber syndrome 1); Eng; HHT1; ORW; ORW1; S-endoglin; transmembrane glycoprotein | |
| ENG | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.