missing translation for 'onlineSavingsMsg'
Learn More

CD10 (Membrane Metalloendopeptidase) Mouse, Clone: MME/1870, enQuireBio™

Mouse Monoclonal Antibody

£181.83 - £423.29

Specifications

Antigen CD10 (Membrane Metalloendopeptidase)
Clone MME/1870
Concentration 200μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15910488
View Documents
EnquireBio
4311-MSM10-P0
20 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15920488
View Documents
EnquireBio
4311-MSM10-P1
100 μg
£423.29
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CD10 (Membrane Metalloendopeptidase)
200μg/mL
Monoclonal
Mouse
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant human CD10 protein fragment (aa297-483)
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
100kDa
MME/1870
Immunohistochemistry
Unconjugated
RUO
4311
IgG2c κ
Protein A/G purified from Bioreactor concentrate.
Cell Surface and Cytoplasmic
Kidney, Prostate or Small Intestine
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.