missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD10 (Membrane Metalloendopeptidase) Mouse, Clone: MME/1870, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 4311-MSM10-P1
This item is not returnable.
View return policy
Specifications
| CD10 (Membrane Metalloendopeptidase) | |
| Cell Surface and Cytoplasmic | |
| MME/1870 | |
| Unconjugated | |
| Mouse | |
| 100kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 4311 | |
| IgG2c κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant human CD10 protein fragment (aa297-483) | |
| Kidney, Prostate or Small Intestine | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction