missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD-M6PR Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£134.00 - £397.00
Specifications
| Antigen | CD-M6PR |
|---|---|
| Concentration | LYOPH |
| Applications | Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18242383
|
Novus Biologicals
NBP1-20167-0.1MG |
0.1 mg |
£134.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18221800
|
Novus Biologicals
NBP1-20167 |
0.5 mg |
£397.00
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD-M6PR Polyclonal antibody specifically detects CD-M6PR in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
| CD-M6PR | |
| Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| P24668 | |
| 4074 | |
| IgG | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| LYOPH | |
| Polyclonal | |
| Purified | |
| RUO | |
| 000 Man6PR, 46 kDa mannose 6-phosphate receptor, 46-kDa mannose 6-phosphate receptor, cation-dependent mannose-6-phosphate receptor, CD Man-6-P receptor, CD-MPR, FLJ32994, M6PR, mannose-6-phosphate receptor (cation dependent), MPR 46, MPR46, MPRD, Mr 46, SMPR | |
| M6PR | |
| Primary | |
| MPRD. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title