missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD-M6PR Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-20167
This item is not returnable.
View return policy
Description
CD-M6PR Polyclonal antibody specifically detects CD-M6PR in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications
| CD-M6PR | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from PBS with No Preservative | |
| 000 Man6PR, 46 kDa mannose 6-phosphate receptor, 46-kDa mannose 6-phosphate receptor, cation-dependent mannose-6-phosphate receptor, CD Man-6-P receptor, CD-MPR, FLJ32994, M6PR, mannose-6-phosphate receptor (cation dependent), MPR 46, MPR46, MPRD, Mr 46, SMPR | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| MPRD. | |
| Human, Mouse, Rat, Primate | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| LYOPH | |
| Western Blot 10-50 ug/ml, Immunohistochemistry 10-50 ug/ml, Immunohistochemistry-Paraffin 10-50 ug/ml | |
| P24668 | |
| M6PR | |
| A synthetic peptide from mouse CD-M6PR conjugated to blue carrier protein was used as the antigen. | |
| 0.5 mg | |
| Immunology | |
| 4074 | |
| Reconstitute 0.5 mg size in 0.5 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the 0.1 mg size is provided in reconstituted format. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction