missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCT7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CCT7 |
|---|---|
| Concentration | 0.2 mg/mL |
| Applications | Western Blot, Immunoprecipitation, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16835424
|
Proteintech
15994-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16825424
|
Proteintech
15994-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TPRBK may be involved in the condensation of spindle midzone microtubules, leading to the formation of midbody. Diseases associated with this gene include Cleft Soft Palate.Specifications
| CCT7 | |
| Western Blot, Immunoprecipitation, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Rat, Mouse | |
| P80313, Q99832 | |
| 10574, 12468, 297406 | |
| CCT7 Fusion Protein Ag8868 | |
| Primary | |
| -20°C | |
| CCT7 |
| 0.2 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CCT ETA, CCT7, Ccth, HIV 1 Nef interacting protein, Nip7 1, TCP 1 eta | |
| CCT7 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title