missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCT7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15994-1-AP-20UL
This item is not returnable.
View return policy
Description
TPRBK may be involved in the condensation of spindle midzone microtubules, leading to the formation of midbody. Diseases associated with this gene include Cleft Soft Palate.Specifications
| CCT7 | |
| Polyclonal | |
| Unconjugated | |
| CCT7 | |
| CCT ETA, CCT7, Ccth, HIV 1 Nef interacting protein, Nip7 1, TCP 1 eta | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10574, 12468, 297406 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation, Immunocytochemistry, Immunofluorescence | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P80313, Q99832 | |
| CCT7 | |
| CCT7 Fusion Protein Ag8868 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction