missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR8 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
£175.00 - £386.00
Specifications
| Antigen | CCR8 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500, ELISA 1:50000, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, ELISA |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18289034
|
Novus Biologicals
NB100-709-0.025MG |
0.025 mg |
£175.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18750474
|
Novus Biologicals
NB100-709 |
£386.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
CCR8 Polyclonal specifically detects CCR8 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CCR8 | |
| Western Blot 1:500, ELISA 1:50000, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200 | |
| Polyclonal | |
| Goat | |
| Cytokine Research, GPCR, Immunology, Innate Immunity, Virology Bacteria and Parasites | |
| 10mM KHPO4 and 0.14M NaCl, BSA with 0.1% Sodium Azide | |
| CC chemokine receptor 8, C-C CKR-8, CC-chemokine receptor chemr1, CC-CKR-8, CCR-8, CDw198, CDw198 antigen, chemokine (C-C motif) receptor 8, chemokine (C-C) receptor 8, chemokine (C-C) receptor-like 2, Chemokine receptor-like 1, CKR-L1, CKRL1MGC129966, CMKBR8MGC129973, CMKBRL2C-C chemokine receptor type 8, CY6, GPRCY6, GPR-CY6, TER1CC chemokine receptor CHEMR1 | |
| CCR8 | |
| IgG | |
| Affinity Purified | |
| This recognizes CCR8 (50 kDa). The peptide sequence is < 50 % identical to other mouse chemokine receptors in this region. |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Mouse | |
| P56484 | |
| 1237 | |
| Synthetic peptide: PNVTMTDYYPDFFTAPCDAEFLLRGSM, corresponding to amino acids 7-33 of Mouse CCR8. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title