missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR8 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-709-0.025MG
This item is not returnable.
View return policy
Description
CCR8 Polyclonal specifically detects CCR8 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CCR8 | |
| Polyclonal | |
| Unconjugated | |
| 10mM KHPO4 and 0.14M NaCl, BSA with 0.1% Sodium Azide | |
| CC chemokine receptor 8, C-C CKR-8, CC-chemokine receptor chemr1, CC-CKR-8, CCR-8, CDw198, CDw198 antigen, chemokine (C-C motif) receptor 8, chemokine (C-C) receptor 8, chemokine (C-C) receptor-like 2, Chemokine receptor-like 1, CKR-L1, CKRL1MGC129966, CMKBR8MGC129973, CMKBRL2C-C chemokine receptor type 8, CY6, GPRCY6, GPR-CY6, TER1CC chemokine receptor CHEMR1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This recognizes CCR8 (50 kDa). The peptide sequence is < 50 % identical to other mouse chemokine receptors in this region. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, ELISA | |
| 1.0 mg/mL | |
| Western Blot 1:500, ELISA 1:50000, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200 | |
| P56484 | |
| CCR8 | |
| Synthetic peptide: PNVTMTDYYPDFFTAPCDAEFLLRGSM, corresponding to amino acids 7-33 of Mouse CCR8. | |
| 0.025 mg | |
| Cytokine Research, GPCR, Immunology, Innate Immunity, Virology Bacteria and Parasites | |
| 1237 | |
| Mouse | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur