missing translation for 'onlineSavingsMsg'
Learn More

CCR7 (CD197) Mouse anti-Human, PE, Clone: 2-L1-A, BD Pharmingen™

Mouse Monoclonal Antibody

£129.96 - £226.52

Specifications

Antigen CCR7 (CD197)
Clone 2-L1-A
Applications Flow Cytometry
Classification Monoclonal
Conjugate PE
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17207574
View Documents
BD Biosciences
566742
25 Tests
ONLINE EXCLUSIVE PRICE
£132.00
£129.96 / 25 tests
Save £2.04
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17297564
View Documents
BD Biosciences
566741
100 Tests
ONLINE EXCLUSIVE PRICE
£368.00
£226.52 / 100 tests
Save £141.48
38% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CCR7 (CD197) Monoclonal antibody specifically detects CCR7 (CD197) in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CCR7 (CD197)
Flow Cytometry
PE
RUO
IgG1 κ
Refrigerate (2 to 8°C)
2-L1-A
Monoclonal
Mouse
Human
Primary
CCR7 (CD197)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.