missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR7 (CD197) Mouse anti-Human, PE, Clone: 2-L1-A, BD Pharmingen™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566741
This item is not returnable.
View return policy
Description
CCR7 (CD197) Monoclonal antibody specifically detects CCR7 (CD197) in Human samples. It is validated for Flow Cytometry.
Specifications
| CCR7 (CD197) | |
| Monoclonal | |
| PE | |
| 100 Tests | |
| Primary | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| 2-L1-A | |
| Mouse | |
| RUO | |
| CCR7 (CD197) | |
| Refrigerate (2 to 8°C) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction