missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR7 Antibody (4B7B8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£174.00 - £443.00
Specifications
| Antigen | CCR7 |
|---|---|
| Clone | 4B7B8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100- 1:500, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18281911
|
Novus Biologicals
NBP2-61680 |
0.1 mL |
£443.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18674228
|
Novus Biologicals
NBP2-61680-0.025ml |
0.025 mL |
£174.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR7 Monoclonal specifically detects CCR7 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| CCR7 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| BLR2C-C chemokine receptor type 7, C-C CKR-7, CC-CKR-7, CCR-7, CD197, CD197 antigen, CDw197CC chemokine receptor 7, chemokine (C-C motif) receptor 7, CMKBR7chemokine (C-C) receptor 7, EBI1EBV-induced G protein-coupled receptor 1, EBV-induced G-protein coupled receptor 1, Epstein-Barr virus induced gene 1, Epstein-Barr virus induced G-protein coupled receptor, Epstein-Barr virus-induced G-protein coupled receptor 1, EVI1, lymphocyte-specific G protein-coupled peptide receptor, MIP-3 beta receptor | |
| CCR7 | |
| IgG1 | |
| Protein G purified |
| 4B7B8 | |
| Western Blot 1:100- 1:500, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 1236 | |
| Purified recombinant fragment of human CD197 (AA: extra mix) expressed in E. Coli. | |
| Primary | |
| 42.9 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title