missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR7 Antibody (4B7B8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61680
This item is not returnable.
View return policy
Description
CCR7 Monoclonal specifically detects CCR7 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| CCR7 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100- 1:500, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
| BLR2C-C chemokine receptor type 7, C-C CKR-7, CC-CKR-7, CCR-7, CD197, CD197 antigen, CDw197CC chemokine receptor 7, chemokine (C-C motif) receptor 7, CMKBR7chemokine (C-C) receptor 7, EBI1EBV-induced G protein-coupled receptor 1, EBV-induced G-protein coupled receptor 1, Epstein-Barr virus induced gene 1, Epstein-Barr virus induced G-protein coupled receptor, Epstein-Barr virus-induced G-protein coupled receptor 1, EVI1, lymphocyte-specific G protein-coupled peptide receptor, MIP-3 beta receptor | |
| Mouse | |
| 42.9 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 4B7B8 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CCR7 | |
| Purified recombinant fragment of human CD197 (AA: extra mix) expressed in E. Coli. | |
| Protein G purified | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 1236 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction