missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCR4 Recombinant Rabbit Monoclonal Antibody (5Q9W4)

Rabbit Recombinant Monoclonal Antibody
£147.00 - £363.00
Specifications
| Antigen | CCR4 |
|---|---|
| Clone | 5Q9W4 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Flow Cytometry, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30283346
|
Invitrogen™
MA557947 |
20 μL |
£147.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30283065
![]() |
Invitrogen™
MA557948 |
100 μL |
£363.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: MNPTDIADTT LDESIYSNYY LYESIPKPCT KEGIKAFGEL FLPPLYSLVF VFGLLGNSVV VLVLFKYKRL RSMTDVYLLN LAISDLLFVF SLPFWGYYAA.
CCR4 is a chemokine receptor and is preferentially expressed on type 2 helper T (Th2-type) cells. In contrast to other chemokine receptors, the expression of CCR4 and CCR8 on Th2 cells is transiently increased following TCR and CD28 engagement. Moreover, activated Th1 cells up-regulate CCR4 expression and functional responsiveness to thymus- and activation-regulated chemokine. Chemokines are a group of small (approximately 8 to 14 kD), mostly basic, structurally related molecules that regulate cell trafficking of various types of leukocytes through interactions with a subset of 7-transmembrane, G protein-coupled receptors. Chemokines also play fundamental roles in the development, homeostasis, and function of the immune system, and they have effects on cells of the central nervous system as well as on endothelial cells involved in angiogenesis or angiostasis. Chemokines are divided into 2 major subfamilies, CXC and CC, based on the arrangement of the first 2 of the 4 conserved cysteine residues; the 2 cysteines are separated by a single amino acid in CXC chemokines and are adjacent in CC chemokines.Specifications
| CCR4 | |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P51679, P51680 | |
| 1233, 12773 | |
| A synthetic peptide corresponding to a sequence within amino acids 16-32 of human CD194/CCR4 (NP_005499.1). | |
| Antibody |
| 5Q9W4 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| Ccr4 | |
| C-C chemokine receptor 4; C-C chemokine receptor type 4; C-C CKR-4; C-C motif chemokine receptor 4; CC-CKR-4; Ccr4; CCR-4; CD194; chemokine (C-C motif) receptor 4; chemokine (C-C) receptor 4; CHEMR1; ChemR13; CKR4; Cmkbr4; fusin; HGCN:14099; K5-5; LESTR; leukocyte-expressed seven-transmembrane-domain receptor; MGC88293; Sdf1r | |
| Ccr4 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title