missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CCR4 Recombinant Rabbit Monoclonal Antibody (5Q9W4)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557947

 View more versions of this product

Product Code. 30283346

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MNPTDIADTT LDESIYSNYY LYESIPKPCT KEGIKAFGEL FLPPLYSLVF VFGLLGNSVV VLVLFKYKRL RSMTDVYLLN LAISDLLFVF SLPFWGYYAA.

CCR4 is a chemokine receptor and is preferentially expressed on type 2 helper T (Th2-type) cells. In contrast to other chemokine receptors, the expression of CCR4 and CCR8 on Th2 cells is transiently increased following TCR and CD28 engagement. Moreover, activated Th1 cells up-regulate CCR4 expression and functional responsiveness to thymus- and activation-regulated chemokine. Chemokines are a group of small (approximately 8 to 14 kD), mostly basic, structurally related molecules that regulate cell trafficking of various types of leukocytes through interactions with a subset of 7-transmembrane, G protein-coupled receptors. Chemokines also play fundamental roles in the development, homeostasis, and function of the immune system, and they have effects on cells of the central nervous system as well as on endothelial cells involved in angiogenesis or angiostasis. Chemokines are divided into 2 major subfamilies, CXC and CC, based on the arrangement of the first 2 of the 4 conserved cysteine residues; the 2 cysteines are separated by a single amino acid in CXC chemokines and are adjacent in CC chemokines.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CCR4
Recombinant Monoclonal
1 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
P51679, P51680
Ccr4
A synthetic peptide corresponding to a sequence within amino acids 16-32 of human CD194/CCR4 (NP_005499.1).
20 μL
Primary
Human, Mouse
Antibody
IgG
ELISA, Flow Cytometry, Western Blot
5Q9W4
Unconjugated
Ccr4
C-C chemokine receptor 4; C-C chemokine receptor type 4; C-C CKR-4; C-C motif chemokine receptor 4; CC-CKR-4; Ccr4; CCR-4; CD194; chemokine (C-C motif) receptor 4; chemokine (C-C) receptor 4; CHEMR1; ChemR13; CKR4; Cmkbr4; fusin; HGCN:14099; K5-5; LESTR; leukocyte-expressed seven-transmembrane-domain receptor; MGC88293; Sdf1r
Rabbit
Protein A
RUO
1233, 12773
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.