missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR4 Goat anti-Human, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody has been used in 6 publications
£141.00
Specifications
| Antigen | CCR4 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500, Flow Cytometry 1:12, ELISA 1:30000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:400, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunohistochemistry, ELISA |
| Classification | Polyclonal |
Description
CCR4 Polyclonal specifically detects CCR4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| CCR4 | |
| Western Blot 1:500, Flow Cytometry 1:12, ELISA 1:30000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:400, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen 1:10-1:500 | |
| Polyclonal | |
| Goat | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 10mM KHPO4, 0.14M NaCl and 1.0 mg/ml BSA. with 0.1% Sodium Azide | |
| C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5 | |
| CCR4 | |
| IgG | |
| Affinity Purified | |
| Peptide sequence is < 50 % identical to other human chemokine receptors in this region. |
| 1.0 mg/mL | |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| RUO | |
| Human | |
| P51679 | |
| 1233 | |
| A synthetic peptide [SNYYLYESIPKPCTKEGIKAFGE] corresponding to amino acids 17-39 of the extracellular domain of human CCR4. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title