missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR4 Goat anti-Human, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NB100-717-0.025MG
This item is not returnable.
View return policy
Description
CCR4 Polyclonal specifically detects CCR4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| CCR4 | |
| Polyclonal | |
| Unconjugated | |
| 10mM KHPO4, 0.14M NaCl and 1.0 mg/ml BSA. with 0.1% Sodium Azide | |
| C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Peptide sequence is < 50 % identical to other human chemokine receptors in this region. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunohistochemistry, ELISA | |
| 1.0 mg/mL | |
| Western Blot 1:500, Flow Cytometry 1:12, ELISA 1:30000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:400, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen 1:10-1:500 | |
| P51679 | |
| CCR4 | |
| A synthetic peptide [SNYYLYESIPKPCTKEGIKAFGE] corresponding to amino acids 17-39 of the extracellular domain of human CCR4. | |
| 0.025 mg | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 1233 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction