missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL8/MCP-2 Antibody (35509.11), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£156.00
Specifications
| Antigen | CCL8/MCP-2 |
|---|---|
| Clone | 35509.11 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1 - 2 μg/mL, ELISA 1 - 2 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:500-1:1000, Immunohistochemistry-Frozen, Neutralization 1 - 4 μg/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18527550
|
Novus Biologicals
NB120-10391-0.025mg |
0.025 mg | |||||||
Description
CCL8/MCP-2 Monoclonal specifically detects CCL8/MCP-2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| CCL8/MCP-2 | |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Chemokines and Cytokines, Cytokine Research, Signal Transduction | |
| C-C motif chemokine 8, chemokine (C-C motif) ligand 8, HC14, MCP2, MCP-2, member 8 (monocyte chemotactic protein 2), monocyte chemoattractant protein 2, SCYA10, small-inducible cytokine A8 | |
| CCL8 | |
| IgG1 | |
| Protein A purified | |
| By ELISA, this antibody shows cross-reactivity with recombinant human MCP1 and recombinat mouse MIP1 alpha. In addition, it does not cross-react with other chemokines tested: rmC10, rhENA78, rhGRO alpha, rhGRO beta, rhGRO gamma, rmJE, rhI309, and rhIL8. This antibody neutralizes the biological activity of recombinant human MCP-2. Shows <20% cross-reactivity with recombinant human MCP-1 and recombinant mouse MIP-1a. |
| 35509.11 | |
| Western Blot 1 - 2 μg/mL, ELISA 1 - 2 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:500-1:1000, Immunohistochemistry-Frozen, Neutralization 1 - 4 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 6355 | |
| Recombinant human monocyte chemotactic protein 2 (MCP2), expressed in E. coli. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title