missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL8/MCP-2 Antibody (35509.11), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB120-10391-0.025mg
This item is not returnable.
View return policy
Description
CCL8/MCP-2 Monoclonal specifically detects CCL8/MCP-2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| CCL8/MCP-2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 1 - 2 μg/mL, ELISA 1 - 2 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:500-1:1000, Immunohistochemistry-Frozen, Neutralization 1 - 4 μg/mL | |
| CCL8 | |
| Recombinant human monocyte chemotactic protein 2 (MCP2), expressed in E. coli. | |
| 0.025 mg | |
| Chemokines and Cytokines, Cytokine Research, Signal Transduction | |
| 6355 | |
| Human | |
| Purified |
| Western Blot | |
| 35509.11 | |
| Unconjugated | |
| C-C motif chemokine 8, chemokine (C-C motif) ligand 8, HC14, MCP2, MCP-2, member 8 (monocyte chemotactic protein 2), monocyte chemoattractant protein 2, SCYA10, small-inducible cytokine A8 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| By ELISA, this antibody shows cross-reactivity with recombinant human MCP1 and recombinat mouse MIP1 alpha. In addition, it does not cross-react with other chemokines tested: rmC10, rhENA78, rhGRO alpha, rhGRO beta, rhGRO gamma, rmJE, rhI309, and rhIL8. This antibody neutralizes the biological activity of recombinant human MCP-2. Shows <20% cross-reactivity with recombinant human MCP-1 and recombinant mouse MIP-1a. | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction