Learn More
Invitrogen™ CCL6 Mouse ELISA Kit
Sandwich ELISA Kit
£504.00 - £2320.00
Specifications
Accession Number | P27784 |
---|---|
Assay Sensitivity | 18 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15485233
|
Invitrogen™
EMCCL6 |
96 Tests |
£504.00
1 set |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
17135699
|
Invitrogen™
EMCCL6X5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Description
The Mouse CCL6 ELISA quantitates Ms CCL6 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms CCL6. Principle of the method The Mouse CCL6 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
CCL6(22-95) and CCL6(23-95) are potent chemoattractants.Specifications
P27784 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Detect mouse CCL6, no cross reactivity with recombinant mouse CCL3, CCL4, or CCL9 | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
20305 | |
<12% | |
HRP | |
Immunology | |
RP23-320E6.4, MRP-1, Scya6, c10 | |
1 hr. 20 min. |
18 pg/mL | |
Biotin | |
Chemokine & Receptors | |
Mouse | |
Colorimetric Microplate Reader | |
MRP-1,Scya6,c10 | |
<10% | |
RUO | |
2°C to 8°C | |
Mouse | |
4 hr. 45 min. |