missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse CCL6 ELISA Kit

Product Code. 15485233
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
1 set
Each
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15485233 96 Tests 1 set
17135699 5 x 96 Tests Each
18527874 10 x 96 Tests Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15485233 Supplier Invitrogen™ Supplier No. EMCCL6

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

The Mouse CCL6 ELISA quantitates Ms CCL6 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms CCL6. Principle of the method The Mouse CCL6 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Chemokine(C-C motif) ligand 6(CCL6) is a small cytokine belonging to the CC chemokine family that has only been identified in rodents. In mice, CCL6 is expressed in cells from neutrophil and macrophage lineages, and can be greatly induced under conditions suitable for myeloid cell differentiation. It is highly expressed in bone marrow cultures that have been stimulated with the cytokine GM-CSF. Some low levels of gene expression also occur in certain cell lines of myeloid origin(e.g. the immature myeloid cell lines DA3 and 32D cl3, and the macrophage cell line P388D) that can also be greatly induced in culture with GM-CSF. However, in activated T cell lines, expression of CCL6 is greatly reduced. CCL6 can also be induced in the mouse lung by the cytokine interleukin 13. Mouse CCL6 is located on chromosome 11. The cell surface receptor for CCL6 is believed to be the chemokine receptor CCR1.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P27784
Assay Range 15.63 to 1000 pg/mL
Assay Sensitivity 18 pg/mL
Calibration Range 18 pg/mL to 1000 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Product Type ELISA
Protein Family Chemokine & Receptors
Sample Type Plasma, Serum, Supernatant
Specificity Detect mouse CCL6, no cross reactivity with recombinant mouse CCL3, CCL4, or CCL9
Sufficient For 96 Assays
Target Species Mouse
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 20305
Gene Symbol MRP-1,Scya6,c10
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Immunology
Sample Volume Plasma 33 μL, Serum 33 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Synonym RP23-320E6.4, MRP-1, Scya6, c10
Target CCL6
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.