missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC170 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £470.00
Specifications
| Antigen | CCDC170 |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/mL, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18684356
|
Novus Biologicals
NBP2-37860-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18699895
|
Novus Biologicals
NBP2-37860 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCDC170 Polyclonal antibody specifically detects CCDC170 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| CCDC170 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q8IYT3 | |
| 80129 | |
| IgG | |
| Immunogen affinity purified |
| Western Blot 0.04-0.4 μg/mL, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol | |
| bA282P11.1, C6orf97, chromosome 6 open reading frame 97, coiled-coil domain containing 170, FLJ23305, hypothetical protein LOC80129 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: RMVSQLEAQISELVEQLGKESGFHQKALQRAQKAENMLETLQGQLTHLEAELVSGGVLRDNLNFEKQKYLKFLDQLSQKMKLDQMA | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title