missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC144NL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | CCDC144NL |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
CCDC144NL Polyclonal specifically detects CCDC144NL in Human samples. It is validated for Western Blot.Specifications
| CCDC144NL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| coiled-coil domain containing 144 family, N-terminal like, MGC87631, putative coiled-coil domain-containing protein 144 N-terminal-like | |
| CCDC144NL | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q6NUI1 | |
| 339184 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title