missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC144NL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57054
This item is not returnable.
View return policy
Description
CCDC144NL Polyclonal specifically detects CCDC144NL in Human samples. It is validated for Western Blot.
Specifications
| CCDC144NL | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| coiled-coil domain containing 144 family, N-terminal like, MGC87631, putative coiled-coil domain-containing protein 144 N-terminal-like | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 339184 | |
| Human | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q6NUI1 | |
| CCDC144NL | |
| Synthetic peptides corresponding to MGC87631 (coiled-coil domain containing 144 family, N-terminal like) The peptide sequence was selected from the middle region of MGC87631. Peptide sequence VDKRDRKKSIQQLVPEYKEKQTPESLPQNNNPAAPSQAEGGEGGVACGTV The peptide sequence for this immunogen was taken from within the described region. | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction