missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBX6 Antibody (8D10A3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £435.00
Specifications
| Antigen | CBX6 |
|---|---|
| Clone | 8D10A3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18138289
|
Novus Biologicals
NBP2-52435 |
0.1 mg |
£435.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18642938
|
Novus Biologicals
NBP2-52435-0.025mg |
0.025 mg |
£161.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CBX6 Monoclonal specifically detects CBX6 in Human samples. It is validated for Western Blot, ELISA, CyTOF-ready.Specifications
| CBX6 | |
| 1 mg/mL | |
| Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| chromobox homolog 6, chromobox protein homolog 6 | |
| CBX6 | |
| IgG1 | |
| Protein A or G purified |
| 8D10A3 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 23466 | |
| Purified recombinant fragment of human CBX6 (AA: 269-412) expressed in E. Coli. | |
| Primary | |
| 43.9 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title