missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBX6 Antibody (8D10A3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52435
This item is not returnable.
View return policy
Description
CBX6 Monoclonal specifically detects CBX6 in Human samples. It is validated for Western Blot, ELISA, CyTOF-ready.
Specifications
| CBX6 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready | |
| chromobox homolog 6, chromobox protein homolog 6 | |
| Mouse | |
| 43.9 kDa | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry | |
| 8D10A3 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CBX6 | |
| Purified recombinant fragment of human CBX6 (AA: 269-412) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| 23466 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction