missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cbx2 Antibody (24A2), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | Cbx2 |
|---|---|
| Clone | 24A2 |
| Dilution | Western Blot 1:500-1:2000, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043179
|
Novus Biologicals
NBP3-26694-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043683
|
Novus Biologicals
NBP3-26694-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cbx2 Monoclonal antibody specifically detects Cbx2 in Human, Rat samples. It is validated for Western Blot, ELISASpecifications
| Cbx2 | |
| Western Blot 1:500-1:2000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| CDCA6, cell division cycle associated 6, chromobox homolog 2, chromobox homolog 2 (Drosophila Pc class), chromobox homolog 2 (Pc class homolog, Drosophila), chromobox protein homolog 2, M33, MGC10561, modifier 3, Pc class homolog | |
| A synthesized peptide derived from Human Cbx2 [UniProt Q14781] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 24A2 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Chromatin Modifiers | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 84733 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title