missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cbx2 Antibody (24A2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26694-50ul
This item is not returnable.
View return policy
Description
Cbx2 Monoclonal antibody specifically detects Cbx2 in Human, Rat samples. It is validated for Western Blot, ELISA
Specifications
| Cbx2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, ELISA | |
| 24A2 | |
| Western Blot 1:500-1:2000, ELISA | |
| CDCA6, cell division cycle associated 6, chromobox homolog 2, chromobox homolog 2 (Drosophila Pc class), chromobox homolog 2 (Pc class homolog, Drosophila), chromobox protein homolog 2, M33, MGC10561, modifier 3, Pc class homolog | |
| A synthesized peptide derived from Human Cbx2 [UniProt Q14781] | |
| 50 μL | |
| Chromatin Modifiers | |
| 84733 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction