missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CBR1 Recombinant Rabbit Monoclonal Antibody (24GB3190)

Rabbit Recombinant Monoclonal Antibody
£126.00 - £434.00
Specifications
| Antigen | CBR1 |
|---|---|
| Clone | 24GB3190 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275389
![]() |
Invitrogen™
MA556999 |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30276399
|
Invitrogen™
MA557000 |
20 μL |
£126.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CBR1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Carbonyl reductase is one of several monomeric, NADPH-dependent oxidoreductases having wide specificity for carbonyl compounds. This enzyme is widely distributed in human tissues. Another carbonyl reductase gene, CRB3, lies close to this gene on chromosome 21q.Specifications
| CBR1 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Rat | |
| CBR1 | |
| 15-hydroxyprostaglandin dehydrogenase; 15-hydroxyprostaglandin dehydrogenase [NADP(+)]; 20-beta-hydroxysteroid dehydrogenase; AW261796; carbonyl reductase (NADPH) 1; carbonyl reductase [NADPH] 1; Carbonyl reductase [NADPH] 1 (NADPH-dependent carbonyl reductase 1); carbonyl reductase 1; CBR; Cbr 1; Cbr1; CR; CRN; hCBR1; NADPH-dependent carbonyl reductase 1; prostaglandin 9-ketoreductase; Prostaglandin-E(2) 9-reductase; SDR21C1; Short chain dehydrogenase/reductase family 21C member 1; short chain dehydrogenase/reductase family 21C, member 1 | |
| CBR1 | |
| Primary | |
| Affinity chromatography |
| 24GB3190 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P16152, P47727 | |
| 29224, 873 | |
| A synthesized peptide derived from human CBR1 (aa 145-185). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title